Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals
Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals
Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals
Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA17259.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS582-24-1
IUPAC Name2-hydroxy-1-phenylethan-1-one
Molecular FormulaC8H8O2
InChI KeyZWVHTXAYIKBMEE-UHFFFAOYSA-N
SMILESOCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 97.0%