1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
CAS: 1012-72-2 | C14H22 | 190.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17146.065 g
Catalog number ALFA17146.06
Price (MYR)
401.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
401.00
Add to cart
Specifications
Chemical Name or Material1,4-Di-tert-butylbenzene
CAS1012-72-2
Melting Point75°C to 79°C
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 9; Packaging Group: III
View more
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.
Solubility
Insoluble in water. Soluble in ethanol, benzene, carbon tetrachloride.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. GH Penner.; JM Polson.; C Stuart. Deuterium NMR and x-ray crystallographic studies of guest and host motions in the thiourea/1,4-di-tert-butylbenzene inclusion compound. J. Phys. Chem. 199296 (12), 5121-5129.
  2. N Gart.; CL Leci.; S Sarig. The effect of solvents on crystal habit of 1, 4-di-tert-butylbenzene (DTBB). Journal of Crystal Growth. 198154 (2), 227-231.