2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
CAS: 104-76-7 | C8H18O | 130.231 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17104.AP500 mL
Catalog number ALFA17104.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS298-81-7
IUPAC Name9-methoxy-7H-furo[3,2-g]chromen-7-one
Molecular FormulaC12H8O4
InChI KeyQXKHYNVANLEOEG-UHFFFAOYSA-N
SMILESCOC1=C2OC=CC2=CC2=C1OC(=O)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point143°C to 148°C
Appearance (Form)Powder or crystals or needles
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Color)White to cream
View more
2-Ethyl-1-hexanol is used as a flavor, fragrance and plasticizer. It is used to prepare diesters bis(2-ethylhexyl) phthalate. It reacts with nitric acid and used as an octane booster. Its ester, 2-ethylhexyl ester is a component of sunscreen octocrylene. Further, it is used as a low volatility solvent for resins, animal fats, waxes, vegetable oils and petroleum derivatives. In addition to this, it is used in plasticizer, dioctyl phthalate, which is used in the production of polyvinyl chloride products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethyl-1-hexanol is used as a flavor, fragrance and plasticizer. It is used to prepare diesters bis(2-ethylhexyl) phthalate. It reacts with nitric acid and used as an octane booster. Its ester, 2-ethylhexyl ester is a component of sunscreen octocrylene. Further, it is used as a low volatility solvent for resins, animal fats, waxes, vegetable oils and petroleum derivatives. In addition to this, it is used in plasticizer, dioctyl phthalate, which is used in the production of polyvinyl chloride products.
Solubility
Miscible with common organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, alkalies, strong bases and strong acids.
RUO – Research Use Only
General References:
  1. Gilani, A. G.; Ansari, M. Dielectric Study of Intermolecular Interactions of 1,3-Propanediol with 1-Octanol and 2-Ethyl-1-hexanol at Various Temperatures. J. Solution Chem. 2014, 43 (8), 1344-1359.
  2. Gilani, A. G.; Gilani, H. G.; Saadat, S. L. S. Solubility and tie line data for the aqueous solutions of butyric acid with 1-octanol and 2-ethyl-1-hexanol at various temperatures. Fluid Phase Equilib. 2014, 361, 45-53.