Bromocresol Green
Bromocresol Green
Bromocresol Green
Thermo Scientific Chemicals
Bromocresol Green
CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA17090.14
Price (MYR)
1,058.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,058.00
Add to cart
Specifications
Chemical Name or MaterialBromocresol Green
Melting Point∼225°C (decomposition)
CAS76-60-8
Recommended StorageAmbient temperatures
RTECS NumberSJ7456000
View more
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
Solubility
Soluble in benzene, ethanol and diethyl ether. Sparingly soluble in water.
Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue).Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ouyang, X.; Luo, L.; Ding, Y.; Liu, B.; Xu, D.; Huang, A. Simultaneous determination of uric acid, dopamine and ascorbic acid based on poly(bromocresol green) modified glassy carbon electrode. J. Electroanal. Chem. 2015, 748, 1-7.
  2. Nezamzadeh-Ejhieh, A.; Moazzeni, N. Sunlight photodecolorization of a mixture of Methyl Orange and Bromocresol Green by CuS incorporated in a clinoptilolite zeolite as a heterogeneous catalyst. J. Ind. Eng. Chem. 2013, 19 (5), 1433-1442.