DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals
CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA17039.30
Price (MYR)
1,238.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
1,238.00
Add to cart
Specifications
Chemical Name or MaterialDL-α-Tocopherol
CAS10191-41-0
Health Hazard 1H317
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c
Melting Point2°C to 4°C
View more
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.
Solubility
Miscible with chloroform, vegetable oils,  ether, acetone and alcohol. Immiscible with water.
Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cimadevilla, H. M.; Hevia, D.; Miar, A.; Mayo, J. C.; Lombo, F.; Sainz, R. M. Development and validation of a single HPLC method for determination of alfa-tocopherol in cell culture and in human or mouse biological samples. Biomed. Chromatogr. 2015, 29 (6), 843-852.
  2. Powell, C. D.; Chowdhury, M. K.; Bureau, D. P. Assessing the bioavailability of l-lysine sulfate compared to l-lysine HCl in rainbow trout (Oncorhynchus mykiss). Aquacult. 2015, 448, (1), 327-333.