2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals
2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals
2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals
2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals
CAS: 88-09-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16989.AK250 mL
Catalog number ALFA16989.AK
Price (MYR)
576.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Specifications
Chemical Name or Material2-Ethylbutyric acid
CAS88-09-5
Health Hazard 1H227-H312-H314
Health Hazard 2GHS H Statement
H311-H314-H318-H227
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.
Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
  1. P.E. Keane.; J. Simiand.; E. Mendes.; V. Santucci.; M. Morre. The effects of analogues of valproic acid on seizures induced by pentylenetetrazol and GABA content in brain of mice.Neuropharmacology,1983,22(7), 875-879.
  2. Hiroshi Miwa.; Magobei Yamamoto. Liquid chromatographic determination of free and total fatty acids in milk and milk products as their 2-nitrophenylhydrazides.J. Chromatogr. A,1990,523,235-246.