Adenine sulfate, 98+%
Adenine sulfate, 98+%
Adenine sulfate, 98+%
Adenine sulfate, 98+%
Thermo Scientific Chemicals
Adenine sulfate, 98+%
CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16964.0910 g
Catalog number ALFA16964.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1784-03-8
IUPAC Namehydrogen methyl 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoate chloride
Molecular FormulaC14H23ClN4O4S
InChI KeyJIQFFACVQXXHMY-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point146°C to 149°C
Infrared spectrumConforms
Titration with NaOH98.0 to 102.0 %
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
  1. Nuki, G. Human purine metabolism: Some recent advances and relationships with immunodeficiency.Ann Rheum Dis.1983,428-11.
  2. Sonali Jana.; Gyan Singh Shekhawat. Plant growth regulators, adenine sulfate and carbohydrates regulate organogenesis and in vitro flowering of Anethum graveolens.Acta Physiologiae Plantarum,2011,33,(2), 305-311.