Xylitol, 99%
Xylitol, 99%
Xylitol, 99%
Thermo Scientific Chemicals
Xylitol, 99%
CAS: 87-99-0 | C5H12O5 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16944.22100 g
Catalog number ALFA16944.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialXylitol
CAS87-99-0
Melting Point92°C to 96°C
Recommended StorageAmbient temperatures
Density1.515 g/mL
View more
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.
Solubility
Soluble in water (50 mg/ml), abs methanol (6 g/100ml), abs ethanol (1.2 g/100ml), alcohol (very slightly), and pyridine. Insoluble in ether.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas.
RUO – Research Use Only
General References:
  1. Amir Azarpazhooh.; Hardy Limeback.; Herenia P Lawrence.; Prakeshkumar S Shah. Xylitol for preventing acute otitis media in children up to 12 years of age.Cochrane Database Syst Rev.2011,11.
  2. Trahan L. Xylitol: a review of its action on mutans streptococci and dental plaque--its clinical significance.International Dental Journal,1995,45.(1), 77-92.