4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 51388-20-6 | C13H14ClNO | 235.711 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16856.1425 g
Catalog number ALFA16856.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1390-65-4
IUPAC Name3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular FormulaC22H20O13
InChI KeyDGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILESCC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to purple
Assay>=42.0 % (Carminic acid)
Infrared spectrumConforms
Loss on drying=<20 %
Appearance (Form)Powder