3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals
CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA16729.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1825-61-2
IUPAC Namemethoxytrimethylsilane
Molecular FormulaC4H12OSi
InChI KeyPOPACFLNWGUDSR-UHFFFAOYSA-N
SMILESCO[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3660 to 1.3680 (20°C, 589 nm)
GC>=96.0 %
Appearance (Color)Clear colorless
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
RUO – Research Use Only
General References:
  1. A Markowska et. al. Synthetic analogues of netropsin and distamycin. IV. Synthesis of a new carbocyclic analogue of distamycin with alkylating side groups.. Journal name. year of publication , 42 (1),129-140 .
  2. S Wongyai et. al. HPLC analysis of polyamines and their acetylated derivatives in the picomole range using benzoyl chloride and 3,5-dinitrobenzoyl chloride as derivatizing agent.. Biomedical Chromatography. 1988, 2(6),254-257.
  3. Reagent for preparation of crystalline derivatives of alcohols: J. Org. Chem., 51, 4819, 4991 (1986); 52, 2086 (1987). Also used for characterization and separation of amines as their sulfonamides: J. Org. Chem., 49, 3043 (1984); 51, 4991 (1986); J. Am. Chem. Soc., 108, 6054 (1986).