2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dithiobiurea, 97%, Thermo Scientific Chemicals
CAS: 142-46-1 | C2H6N4S2 | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA16667.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS511-67-1
IUPAC Name5,5-dibromo-1,3-diazinane-2,4,6-trione
Molecular FormulaC4H2Br2N2O3
InChI KeyAMATXUCYHHHHHB-UHFFFAOYSA-N
SMILESBrC1(Br)C(=O)NC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=95.0 %
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C)
Appearance (Color)White to almost white
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths
Solubility
Soluble in hot water, pyridine, triethylamine and dimethylformamide. Insoluble in cold water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Safin, D. A.; Babashkina, M. G.; Bolte, M.; Mitoraj, M. P.; Klein, A. Metal ion influences distortion of the ligand in the structure of [M{2-MeO(O)CC6H4NHC(S)NP(S)(OiPr)2}2] (M = ZnII, CdII) complexes: a driving force for intermolecular aggregation. Dalton Trans. 2015, 44, 14101-14109.
  2. Yan, Z.; Sun, B.; Guo, C.; Li, Y. Synthesis and properties of azothiazole based pi-conjugated polymers. J. Mater. Chem. C 2014, 2 (34), 7096-7103.