Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals
Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals
Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals
Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals
CAS: 565-73-1 | C2H2NNaO3 | 111.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA16532.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.005 %
Calcium (Ca)=<0.005 %
Copper (Cu)=<0.002 %
Lead (Pb)=<0.002 %
Appearance (Color)Orange to red to pink-brown to red-brown
View more
Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.
Solubility
Soluble in water (10 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Phedias Diamandis et. al.Chemical genetics reveals a complex functional ground state of neural stem cells. Nature Chemical Biology. 2007, 3(5), 268-273 .
  2. G.N.R. Tripathi,; J.E. Katon.Vibrational spectra and structure of crystalline oxamic acid and sodium oxamate . Spectrochimica Acta Part A: Molecular Spectroscopy. 1979, 35 (5), 401-407.