3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16502.30250 g
Catalog number ALFA16502.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS111-87-5
IUPAC Nameoctan-1-ol
Molecular FormulaC8H18O
InChI KeyKBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILESCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
RUO – Research Use Only
General References:
  1. VR Pedireddi.; S Varughese. Solvent-dependent coordination polymers: Cobalt complexes of 3, 5-dinitrobenzoic acid and3, 5-dinitro-4-methylbenzoic acid with 4, 4'-bipyrdine. Inorg. Chem. 2004, 43, (2), 450-457.
  2. JL Lopez.; FSG Einschlag.; MC González. Hydroxyl radical initiated photodegradation of 4-chloro-3, 5-dinitrobenzoic acid in aqueous solution. Journal of Photochemistry and Photobiology . 2000, 137, (2-3), 177-184.