Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals
Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals
Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals
Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals
CAS: 827-52-1 | C12H16 | 160.26 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA16320.22
Catalog number ALFA16320.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight160.26
Unit Size
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Huirache-Acuna, R.; Alonso-Núñez, G.; Paraguay-Delgado, F.; Lara-Romero, J.; Berhault, G.; Rivera-Muñoz, E. M. Unsupported trimetallic CoMoW sulfide HDS catalysts prepared by in situ decomposition of sulfur-containing precursors. Catal. Today 2015, 250, 28-37.
  2. Schultz, C.; Kraft, V.; Pyschik, M.; Weber, S.; Schappacher, F.; Winter, M.; Nowak, S. Separation and Quantification of Organic Electrolyte Components in Lithium-Ion Batteries via a Developed HPLC Method. J. Electrochem. Soc. 2015, 162 (4), A629-A634.