1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
CAS: 106-46-7 | C6H4Cl2 | 146.998 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA16290.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS393-06-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
Water Content (Karl Fischer Titration)≤1.5%
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
RUO – Research Use Only
General References:
  1. M Charbonneau.; J Strasser.; EA Lock.; MJ Turner. Involvement of reversible binding to α 2u-globulin in 1, 4-dichlorobenzene-induced nephrotoxicity. Toxicology and Applied Pharmacology. 1989, 99, (1), 122-132.
  2. RR Jones.; RG Bergman. p-Benzyne. Generation as an intermediate in a thermal isomerization reaction and trapping evidence for the 1, 4-benzenediyl structure. J. Am. Chem. Soc. 1972, 94, (2), 660-661.