Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Actual product may vary
Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals
CAS: 10196-18-6 | N{2}O{6}Zn^0.6 H{2}O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA16282.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Pointca 36°C
CAS10196-18-6
Physical FormSolid
Quantity500 g
Molecular FormulaN{2}O{6}Zn^0.6 H{2}O
Percent Purity98%
Unit Size