Magnesium stearate, Thermo Scientific Chemicals
Magnesium stearate, Thermo Scientific Chemicals
Magnesium stearate, Thermo Scientific Chemicals
Magnesium stearate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium stearate, Thermo Scientific Chemicals
CAS: 557-04-0 | C36H70MgO4 | 591.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA16244.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7782-86-7
IUPAC Namemercury(1+) hydrate nitrate
Molecular FormulaH2HgNO4
InChI KeyUPCSQZXLTBTBCU-UHFFFAOYSA-N
SMILESO.[Hg+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay contain small amounts of dark particulate matter as part of manufacturing process
Appearance (Color)White to yellow to yellow-red
FormCrystalline or lumpy powder
Assay (unspecified)≥96.0%
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
RUO – Research Use Only
General References:
  1. Lebeau, D.; Beuvier, L.; Cornaton, M.; Miserque, F.; Tabarant, M.; Esnouf, S.; Ferry, M. Aging of magnesium stearate under high doses gamma irradiation and oxidative conditions. J. Nucl. Mater. 2015, 460, 130-138.
  2. Haware, R. V.; Shivagari, R.; Johnson, P. R.; Staton, S.; Stagner, W. C.; Gupta, M. R. Application of Multivariate Methods to Evaluate the Functionality of Bovine- and Vegetable-Derived Magnesium Stearate. J. Pharm. Sci. 2014, 103 (5), 1466-1477.