Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, flake, 85%, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16199.0E2500 g
Catalog number ALFA16199.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS22817-26-1
IUPAC Name3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Molecular FormulaC12H11N
InChI KeyNBWCGDRKSOXZNI-UHFFFAOYSA-N
SMILESC1NCC2=C3C1=CC=CC3=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)96.0-105.0?C
Appearance (Color)Green to brown
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.
Solubility
Freely soluble in water. Soluble in ethanol.
Notes
Incompatible with acids, nitro compounds, alkali metals, magnesium, copper, halogens, azides and anhydrides. Contact with aluminum, tin and zinc liberates hydrogen gas.
RUO – Research Use Only
General References:
  1. Aili, D.; Jankova, K.; Li, Q.; Bjerrum, N. J.; Jensen, J. O. The stability of poly(2,2'-(m-phenylene)-5,5'-bibenzimidazole) membranes in aqueous potassium hydroxide. J. Membr. Sci. 2015, 492, 422-429.
  2. Gilliama, R. J.; Graydonb, J. W.; Kirkb, D. W.; Thorpea, S. J. A Review of Specific Conductivities of Potassium hydroxide Solutions for Various Concentrations and Temperatures. Int. J. Hydrogen Energ. 2007, 32 (3), 359-364.
  3. Baroutian, S.; Aroua, M. K.; Raman, A. A. A.; Sulaiman, N. M. N. Potassium Hydroxide Catalyst Supported on Palm Shell Activated Carbon for Transesterification of Palm Oil. Fuel Process. Technol. 2010, 91 (11), 1378-1385.