5-Bromo-2-fluorobenzonitrile, 98%, Thermo Scientific Chemicals
5-Bromo-2-fluorobenzonitrile, 98%, Thermo Scientific Chemicals
5-Bromo-2-fluorobenzonitrile, 98%, Thermo Scientific Chemicals
5-Bromo-2-fluorobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-2-fluorobenzonitrile, 98%, Thermo Scientific Chemicals
CAS: 179897-89-3 | C7H3BrFN | 200.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16113.065 g
Catalog number ALFA16113.06
Price (MYR)
296.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
296.00
Add to cart
Specifications
Chemical Name or Material5-Bromo-2-fluorobenzonitrile
CAS179897-89-3
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c