4-Chloro-2-fluorobenzaldehyde, 98%
4-Chloro-2-fluorobenzaldehyde, 98%
Thermo Scientific Chemicals
4-Chloro-2-fluorobenzaldehyde, 98%
Have Questions?
Catalog NumberQuantity
ALFA16110.065 g
Catalog number ALFA16110.06
Price (MYR)
415.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Specifications
Melting Point58°C to 60°C
Quantity5 g
Percent Purity98%
Physical FormSolid
Unit Size
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.