Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals
Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals
Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals
Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA16068.AU1000 mL
Catalog number ALFA16068.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol mono-n-butyl ether
Melting Point-68°C
CAS112-34-5
Health Hazard 1H319-H373
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
View more
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.
Solubility
Miscible with water, ethyl ether, ethanol acetone and benzene.
Notes
Store in cool place. Incompatible with strong oxidizing agents and light metals.
RUO – Research Use Only
General References:
  1. Wakoh, H.; Furuie, M.; Inaba, D.; Azuma, N.; Nakane, K.; Ogata, N.; Shimizu, T.; Ishimaru, O. Deep-colour vat dyeing of cotton knit fabric on a modified jet dyeing machine. Color. Technol. 2015, 131 (2), 136-141.
  2. Liu, L.; Kim, G. Y.; Hillier, A. C.; Chandra, A. Microstructural and electrochemical impedance study of nickel-Ce0.9Gd0.1O1.95 anodes for solid oxide fuel cells fabricated by ultrasonic spray pyrolysis. J. Power Sources 2011, 196 (6), 3026-3032.