Triton X-100, Thermo Scientific Chemicals
Triton X-100, Thermo Scientific Chemicals
Triton X-100, Thermo Scientific Chemicals
Triton X-100, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triton X-100, Thermo Scientific Chemicals
CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA16046.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS25152-84-5
IUPAC Name(2E,4E)-deca-2,4-dienal
Molecular FormulaC10H16O
InChI KeyJZQKTMZYLHNFPL-BLHCBFLLSA-N
SMILESCCCCC\C=C\C=C\C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=92 % (main isomer)
Refractive index1.5140 to 1.5180 (20°C, 589 nm)
GC>=94 % (sum of isomers)
Appearance (Color)Clear yellow
Appearance (Form)Liquid
View more
Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Nonionic detergent and emulsifier.
  2. El-Agamey, A.; El-Hagrasy, M. A.; Suenobu, T.; Fukuzumi, S. Influence of pH on the decay of beta-carotene radical cation in aqueous Triton X-100: A laser flash photolysis study. J. Photochem. Photobiol., B 2015, 146, 68-73.
  3. Patel, V.; Bharatiya, B.; Ray, D.; Aswal, V. K.; Bahadur, P. Investigations on microstructural changes in pH responsive mixed micelles of Triton X-100 and bile salt. J. Colloid Interface Sci. 2015, 441, 106-112.