Sodium hydroxide, pellets, 98%
Sodium hydroxide, pellets, 98%
Sodium hydroxide, pellets, 98%
Sodium hydroxide, pellets, 98%
Sodium hydroxide, pellets, 98%
Thermo Scientific Chemicals
Sodium hydroxide, pellets, 98%
CAS: 1310-73-2
Have Questions?
Catalog NumberQuantity
ALFA16037.36500 g
Catalog number ALFA16037.36
Price (MYR)
341.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
341.00
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
pH14
Boiling Point1390°C
CAS1310-73-2
Health Hazard 1H314-H335
View more
Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of
RUO – Research Use Only
General References:
  1. Michalska, K.; Bizukojc, M.; Ledakowicz, S. Pretreatment of energy crops with sodium hydroxide and cellulolytic enzymes to increase biogas production. Biomass Bioenergy 2015, 80, 213-221.
  2. Zhu, Q. L.; Tsumori, N.; Xu, Q. Sodium hydroxide-assisted growth of uniform Pd nanoparticles on nanoporous carbon MSC-30 for efficient and complete dehydrogenation of formic acid under ambient conditions. Chem. Sci. 2014, 5 (1), 195-199.