cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
CAS: 585-84-2 | C6H6O6 | 174.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16010.065 g
Catalog number ALFA16010.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialcis-Aconitic acid
CAS585-84-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
View more
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.
Solubility
Soluble in water, alcohol. Slightly soluble in ether.
RUO – Research Use Only
General References:
  1. Mutulis, F.; Kreicberga, J.; Yahorava, S.; Mutule, I.; Borisova-Jan, L.; Yahorau, A.; Muceniece, R.; Azena, S.; Veiksina, S.; Petrovska, R.; Wikberg, J. E. S. Sulfitobacter pseudonitzschiae sp. nov., isolated from the toxic marine diatom Pseudo-nitzschia multiseries. Int. J. Syst. Evol. Microbiol. 2015, 65 (1), 95-100.
  2. Marques, A. C. Q.; Paludo, K. S.; Dallagassa, C. B.; surek, M.; Pedrosa, F. O.; Souza, E. M.; Cruz, L. M.; LiPuma, J. J.; Zanata, S. M.; Rego, F. G. M.; Fadel-Picheth, C. M. T. Biochemical Characteristics, Adhesion, and Cytotoxicity of Environmental and Clinical Isolates ofHerbaspirillum spp. J. Clin. Microbiol. 2015, 53 (1), 302-308.