4-Methoxythiophenol, 97%, Thermo Scientific Chemicals
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALFA16009.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1458-98-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
GC>=96.0 %
StabilizerApprox 500 ppm hydroquinone
Infrared spectrumConforms
View more
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM?s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
RUO – Research Use Only
General References:
  1. Tsung-Chin Tseng.; Ming-Jing Wu. Asymmetric conjugate addition of thiols to chiral methacryloyloxazolidinones. Tetrahedron: Asymmetry. 1995, 6 (7), 1633-1640.
  2. Syed Tasadaque Ali Shah.; Khalid Mohammed Khan.; Angelica Martinez Heinrich.; Wolfgang Voelter. An alternative approach towards the syntheses of thioethers and thioesters using CsF-Celite in acetonitrile. Tetrahedron Letters. 2002, 43 (46), 8281-8283.