Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Thermo Scientific Chemicals
Tetrazolium Violet, 98+%
CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFA16008.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS10099-58-8
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total conc. of imp.as REO=<0.1 %
Appearance (Color)White to off-white