1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 554-95-0 | C9H6O6 | 210.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
250 g
Catalog number ALFA15947.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3,5-Benzenetricarboxylic acid
CAS554-95-0
Health Hazard 1H315-H319-H335-H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.
Solubility
Soluble in water, ethanol and methanol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Choi, Y.; Kim, Y. R.; Kang, Y. S.; Kang, S. W. Enhanced CO2 separation performance of polymer composite membranes through the synergistic effect of 1,3,5-benzenetricarboxylic acid. Chem. Eng. J. 2015, 279, 273-276.
  2. Liu, W.; He, H.; Song, Y.; Du, J.; Sun, F. Solvent-induced generation of two cadmium-based metal-organic frameworks from 1,3,5-benzenetricarboxylic acid ligand. Inorg. Chem. Commun. 2015, 57, 11-14.