N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
CAS: 128-08-5 | C4H4BrNO2 | 177.985 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15922.30250 g
Catalog number ALFA15922.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS639-58-7
IUPAC Nametriphenyltin chloride
Molecular FormulaC18H17ClSn
InChI KeyZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILESCl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥94.0 to ≤106.0%%
CommentAs a bulk commodity material may contain small quantites particulate matter from the manufacturing process
Melting Point (clear melt)103-109°C
View more
N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.
Solubility
Soluble in acetone, tetrahydrofuran, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in water and acetic acid. Insoluble in ether, hexane and carbon tetrachloride.
Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, iron, iron salts, halogenated hydrocarbons, amines and metals. Store in a cool place.
RUO – Research Use Only
General References:
  1. Source of free-radical or positive bromine. For examples of radical benzylic bromination (Wohl-Ziegler), see: Org. Synth. Coll., 4, 921 (1963); 5, 145, 329, 825 (1973). The use of the ozone depleting solvent CCl4 has been avoided by the use of the ionic liquid 1-n-Butyl-3-methyl imidazolium hexafluorophosphate, L19086 : Synlett, 702 (2003). Cleavage of carbohydrate benzyl ethers: J. Org. Chem., 55, 378 (1990), and of benzyl esters: Synlett, 219 (1994), occur under mild conditions; the benzyl group is converted to benzaldehyde. For allylic bromination, see e.g.: Org. Synth. Coll., 4, 108 (1963);.9, 112, 191 (1998) review: Chem. Rev., 43, 271 (1948).
  2. For free-radical ɑ-bromination of a Boc glycine ester, see: Org. Synth. Coll., 9, 526 (1998).
  3. Both aliphatic and aromatic aldehydes are converted to acyl bromides under free-radical conditions: Tetrahedron Lett., 3809 (1979); Synlett, 347 (1990); Tetrahedron Lett., 31, 7237 (1990).
  4. In acetonitrile, NBS is a mild and regiospecific nuclear brominating agent for activated aromatics such as methoxybenzenes and naphthalenes: J. Org. Chem., 60, 5328 (1995). Deactivated aromatics, e.g. nitroarenes and benzotrifluorides, can be m-brominated under mild conditions in good yield with NBS in TFA, in the presence of H2SO4: Synlett, 1245 (1999).
  5. In the presence of a phosphine or phosphite, converts alcohols to alkyl bromides with inversion: Tetrahedron Lett., 3937 (1973). For a review of this and related reactions, see: Org. React., 29, 1 (1983).