S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™
S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™
S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™
S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™
S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA15914.1850 g
Catalog number ALFA15914.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialS-Benzylisothiouronium chloride
CAS538-28-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It has been studied as a small-molecule inhibitor of indoleamine-2,3-dioxygenase.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store in cool dry place. Keep away from oxidizing agent
General References:
  1. Andre S. Dreiding.; Richard J. Pratt. The Carboxylation of cis- and trans-2-Butenyl-2-lithium. A Stereospecific Synthesis of Angelic Acid. J. Am. Chem. Soc. 1954, 76 (7),1902-1906 .
  2. Remsen Ten Eyck.; Isidore Danishefsky. The Addition of Bisulfite to Unsaturated Acids and Their Derivatives.J. Org. Chem. 1951, 16 (11),1683-1689 .