3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 102-32-9 | C8H8O4 | 168.148 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15893.065 g
Catalog number ALFA15893.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS112-24-3
IUPAC Name(2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine
Molecular FormulaC6H18N4
InChI KeyVILCJCGEZXAXTO-UHFFFAOYSA-N
SMILESNCCNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤150
Refractive Index1.4950-1.5010 @20°C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥55.0%
FormLiquid
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air sensitive. Incompatible with strong bases and strong oxidizing agents. Store at room temperature
RUO – Research Use Only
General References:
  1. Ariel Y. Deutch.; See-Ying Tam.; Robert H. Roth. Footshock and conditioned stress increase 3, 4-dihydroxyphenylacetic acid (DOPAC) in the ventral tegmental area but not substantia nigra. Brain Research. 1985, 333 (1), 143-146.
  2. Nicholas G. Bacopoulos.; Susan E. Hattox.; Robert H. Roth. 3,4-Dihydroxyphenylacetic acid and homovanillic acid in rat plasma: Possible indicators of central dopaminergic activity. European Journal of Pharmacology. 1979, 56 (3), 225-236.