Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA15656.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS32005-36-0
IUPAC Namebis((1E,4E)-1,5-diphenylpenta-1,4-dien-3-one) palladium
Molecular FormulaC34H28O2Pd
InChI KeyUKSZBOKPHAQOMP-SVLSSHOZSA-N
SMILES[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Red-brown to black
Assay16 to 21 % (Pd)
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
Solubility
Soluble in water. Slightly soluble ethanol.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Gmitrov, J. Baroreceptor stimulation enhanced nitric oxide vasodilator responsiveness, a new aspect of baroreflex physiology. Microvasc. Res. 2015, 98, 139-144.
  2. Najar, M. H.; Majid, K. Synthesis and characterization of nanocomposite of polythiophene with Na2[Fe(CN)3(OH)(NO)C6H12N4]H2O: a potent material for EMI shielding applications. J. Mater. Sci.- Mater. Electron. 2015, 26 (9), 6458-6470.