4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
Solubility
Soluble in water, alcohol and ether.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
RUO – Research Use Only
General References:
- Zafar A. Siddiqi; Mohd Khalid; Sarvendra Kumar; M. Shahid; Shabana Noor. Antimicrobial and SOD activities of novel transition metal complexes of pyridine-2,6-dicarboxylic acid containing 4-picoline as auxiliary ligand. European Journal of Medicinal Chemistry. 2010,45 (1), 264-269.
- Takeshi Sakamoto; John M. Joern; Akira Arisawa; and Frances H. Arnold. Laboratory Evolution of Toluene Dioxygenase To Accept 4-Picoline as a Substrate. Appl. Environ. Microbiol. 2001, 67 (9), 3882-3887.