Uracil, 99+%
Uracil, 99+%
Uracil, 99+%
Thermo Scientific Chemicals
Uracil, 99+%
CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15570.1850 g
Catalog number ALFA15570.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialUracil
CAS66-22-8
Melting Point∽330°C
Recommended StorageAmbient temperatures
RTECS NumberYQ8650000
View more
Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.
Solubility
Soluble in water, aqueous ammonia, 1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.
Notes
Incompatible with water, air and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. For a review of the use of uracils as starting materials in heterocyclic synthesis, see: Adv. Het. Chem., 55, 130 (1992).
  2. Fornaro, T.; Burini, D.; Biczysko, M.; Barone, V. Hydrogen-Bonding Effects on Infrared Spectra from Anharmonic Computations: Uracil-Water Complexes and Uracil Dimers. J. Phys. Chem. A 2015, 119 (18), 4224-4236.
  3. Cavaleiro, A. M.; Kim, S. H.; Seppala, S.; Nielsen, M. T.; Nørholm, M. H. Accurate DNA assembly and genome engineering with optimized uracil excision cloning. ACS Synth. Biol. 2015, 4 (9), 1042-1046.