1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.
Solubility
Soluble in water 67.2 mg/L.
Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- G M Zaitsev; J S Uotila; I V Tsitko; A G Lobanok and M S Salkinoja-Salonen. Utilization of Halogenated Benzenes, Phenols, and Benzoates by Rhodococcus opacus GM-14. Appl. Environ. Microbiol. 1995, 61 (12), 4191-4201.
- Biao Jiang; Ying Kan; Ao Zhang. Synthesis of dialkyl cyanoboronates and their application in palladium-catalyzed cyanation of aryl halides. Tetrahedron. 2001, 57(8), 1581-1584.