4-Fluorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Fluorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Fluorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Fluorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Fluorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 1544-68-9 | C7H4FNS | 153.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA15561.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1594-56-5
IUPAC Name[(2,4-dinitrophenyl)sulfanyl]formonitrile
Molecular FormulaC7H3N3O4S
InChI KeyXQDQRCRASHAZBA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(SC#N)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Melting Point (clear melt)137.5-143.5?C
Assay (GC)≥97.5%