4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Methoxy-o-phenylenediamine, 98%, Thermo Scientific Chemicals
CAS: 102-51-2 | C7H10N2O | 138.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15557.031 g
Catalog number ALFA15557.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS17084-13-8
IUPAC Namepotassium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6KP
InChI KeyYZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES[K+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
Appearance (Color)White
Water Content (Karl Fischer Titration)<1.0%
FormPowder, crystalline powder or square crystals
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Air sensitive and Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ojeda, A. G.; Wrobel, K.; Escobosa, A. R. C.; Sevilla, E. G.; Wrobel, K. High-performance liquid chromatography determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent. Anal. Biochem. 2014, 449, 52-58.
  2. Vadagaonkar, K. S.; Kalmode, H. P.; Mrugan, K.; Chaskar, A. C. I2 catalyzed tandem protocol for synthesis of quinoxalines via sp3, sp2and sp C-H functionalization. RSC Adv. 2015, 5 (8), 5580-5590.