2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA15412.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.3890 to 1.3910 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Notes
Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Tsilikochrisos, G.; Tsaniklidis, G.; Delis, C.; Nikoloudakis, N.; Aivalakis, G. Glutamate dehydrogenase is differentially regulated in seeded and parthenocarpic tomato fruits during crop development and postharvest storage. Sci. Hortic. 2015, 181, 34-42.
  2. Song, X. Y.; Chen, J.; Shi, Y. P. Electromembrane extraction based on carbon nanotubes reinforced hollow fiber for the determination of plant hormones. New J. Chem. 2015, 39 (12), 9191-9199.