4-Benzylbiphenyl, 98+%, Thermo Scientific Chemicals
4-Benzylbiphenyl, 98+%, Thermo Scientific Chemicals
4-Benzylbiphenyl, 98+%, Thermo Scientific Chemicals
4-Benzylbiphenyl, 98+%, Thermo Scientific Chemicals
4-Benzylbiphenyl, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA15392.065 g
Catalog number ALFA15392.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS12032-20-1
IUPAC Namedilutetium(3+) trioxidandiide
Molecular FormulaLu2O3
InChI KeyUGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Lu+3].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=99.94 %
Appearance (Form)Powder