Coumarin, 98%, Thermo Scientific Chemicals
Coumarin, 98%, Thermo Scientific Chemicals
Coumarin, 98%, Thermo Scientific Chemicals
Coumarin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Coumarin, 98%, Thermo Scientific Chemicals
CAS: 91-64-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA15336.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCoumarin
CAS91-64-5
Health Hazard 1H302+H312+H332-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Nawal Kishor Mal; Masahiro Fujiwara; Yuko Tanaka. Photocontrolled reversible release of guest molecules from coumarin-modified mesoporous silica. Nature. 2003, 421,350-353.
  2. Guilford JonesII; William R.Jackson; Chol Yoo Choi; William R.Bergmark. Solvent effects on emission yield and lifetime for coumarin laser dyes. Requirements for a rotatory decay mechanism. J. Phys. Chem. 1985, 89,(2), 294-300.