2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
CAS: 616-45-5 | C4H7NO | 85.106 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA15332.36
Catalog number ALFA15332.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight85.11
Unit Size
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.
Solubility
Miscible with water and other organic solvents.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only
General References:
  1. Jiang, H.; Yuana, H.; Liang, Y.; Xia, S.; Zhao, Q.; Wu, Q.; Zhang, L.; Lianga, Z.; Zhang, Y. A hydrophilic immobilized trypsin reactor with N-vinyl-2-pyrrolidinone modified polymer microparticles as matrix for highly efficient protein digestion with low peptide residue. J. Chromatogr. A 2012, 1246, 111-116.
  2. Shahbuddin, M.; Bullock, A. J.; MacNeil, S.; Rimme, S. Glucomannan-poly(N-vinyl pyrrolidinone) bicomponent hydrogels for wound healing. J. Mater. Chem. B 2014, 2 (6), 727-738.