1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
CAS: 90-12-0 | C11H10 | 142.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA15327.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Assay from Supplier's CofA>99.9%
FormRod, 7mm (0.28in) diameter (Glassy Carbon type-2, maximimum service temperature 3000?C).
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide
Solubility
Miscible with water, alcohol, ethanol, ether and benzene.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
RUO – Research Use Only
General References:
  1. Onyango, E. O.; Kelley, A. R.; Qian, D. C.; Gribble, G. W. Syntheses of 1-Bromo-8-methylnaphthalene and 1-Bromo-5-methylnaphthalene. J. Org. Chem. 2015, 80 (11), 5970-5972.
  2. Jaroszewska, K.; Masalska, A.; Grzechowiak, J. R.; Grams, J. Hydroconversion of 1-methylnaphthalene over Pt/AlSBA-15-Al2O3 composite catalysts. Appl. Catal., A 2015, 505, 116-130.