4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA15307.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration with HCl>=98.0 %
Heavy metals (as Pb)=<0.001 %
Insoluble matter=<0.005 %
View more
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. O.A. Oyedapo; C.O. Adewunmi; E.O. Iwalewa and V.O. Makanju. Analgesic, Antioxidant and Anti-inflammatory Related Activities of 21-hydroxy-2,41-dimethoxychalcone and 4-hydroxychalcone in Mice.Journal of Biological Sciences.2008, 8 131-136.
  2. Barbora Orlikovaa; Deniz Tasdemirb; Frantisek Golaisc; Mario Dicatoa; Marc Diedericha. The aromatic ketone 4'-hydroxychalcone inhibits TNFα-induced NF-κB activation via proteasome inhibition.Biochemical Pharmacology.2011, 82 620-631.