3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
CAS: 95-76-1 | C6H5Cl2N | 162.013 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15199.1850 g
Catalog number ALFA15199.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS3587-64-2
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.4030-1.4070 @ 20?C