trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
CAS: 501-98-4 | C9H8O3 | 164.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15167.1425 g
Catalog number ALFA15167.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS52-90-4
IUPAC Name2-amino-3-sulfanylpropanoic acid
Molecular FormulaC3H7NO2S
InChI KeyXUJNEKJLAYXESH-UHFFFAOYNA-N
SMILESNC(CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Chloride (Cl)=<0.05 %
Iron (Fe)=<10 ppm
Specific optical rotation+7.5° to +9.5° (20°C, 589 nm) (c=8, 5N HCl)
pH4.5 to 5.5 (2.5 % in water)
View more
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Olga I. Tarzi.; Hiroshi Nonami.; Rosa Erra-Balsells. The effect of temperature on the stability of compounds used as UV-MALDI-MS matrix: 2,5-dihydroxybenzoic acid, 2,4,6-trihydroxyacetophenone, α-cyano-4-hydroxycinnamic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, nor-harmane and harmane. Journal of Mass Spectrometry. 2009, 44, (2), 260-277.
  2. Jeong-Yong Cho.; Jae-Hak Moon.; Ki-Young Seong.; Keun-Hyung Park. Antimicrobial Activity of 4-Hydroxybenzoic Acid and trans 4-Hydroxycinnamic Acid Isolated and Identified from Rice Hull. Bioscience, Biotechnology, and Biochemistry. 1998, 62, (11), 2273-2276.