4-Bromoaniline, 98+%, Thermo Scientific Chemicals
4-Bromoaniline, 98+%, Thermo Scientific Chemicals
4-Bromoaniline, 98+%, Thermo Scientific Chemicals
4-Bromoaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromoaniline, 98+%, Thermo Scientific Chemicals
CAS: 106-40-1 | C6H6BrN | 172.025 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15126.1850 g
Catalog number ALFA15126.18
Price (MYR)
279.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
279.00
Add to cart
Specifications
Chemical Name or Material4-Bromoaniline
CAS106-40-1
Health Hazard 1H302+H332-H311-H315-H319-H373
Health Hazard 2GHS H Statement
H311-H373-H302-H332-H315-H319
Health Hazard 3P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
View more
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.
Solubility
Soluble in alcohol and ether. Insoluble in cold water.
Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Gao, W. Synthesis and Structure Analysis of N-diphenylphosphoryl-4-bromoaniline. Mol. Cryst. Liq. Cryst. 2015, 609 (1), 155-160.
  2. Štrukil, V.; Gracin, D.; Magdysyuk, O. V.; Dinnebier, R. E.; Friščić, T. Trapping Reactive Intermediates by Mechanochemistry: Elusive Aryl N-Thiocarbamoylbenzotriazoles as Bench-Stable Reagents. Angew. Chem. Int. Ed. 2015, 54 (29), 8440-8443.