2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
CAS: 75-30-9 | C3H7I | 169.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA15066.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10581-38-1
IUPAC Name2,2,6,6-tetramethylpiperidin-4-one hydrate
Molecular FormulaC9H19NO2
InChI KeySFDODBGIDPNFEH-UHFFFAOYSA-N
SMILESO.CC1(C)CC(=O)CC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)58.0-62.0?C
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.
Solubility
Soluble in water(1.4g/L).
Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. The ring-opening of cyclic thionocarbonates followed by reductive elimination with zinc to give alkenes provides an alternative to the Corey-Winter desulfurization method (see Triethyl phosphite, L00339): Tetrahedron Lett., 3793 (1973).