2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
CAS: 609-54-1 | C8H10O3S | 186.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALFA15007.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,5-Dimethylbenzenesulfonic acid hydrate
Melting Point84°C to 86°C
CAS609-54-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement