2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimethylbenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals
CAS: 609-54-1 | C8H10O3S | 186.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ALFA15007.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7803-63-6
IUPAC Nameammonium hydrogen sulfate
Molecular FormulaH5NO4S
InChI KeyBIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES[NH4+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration with NaOH98.5 to 101.5 %
Arsenic (As)=<0.00002 %
Cobalt (Co)=<0.0005 %