4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
CAS: 2393-23-9 | C8H11NO | 137.182 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14999.1425 g
Catalog number ALFA14999.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS111-90-0
IUPAC Name2-(2-ethoxyethoxy)ethan-1-ol
Molecular FormulaC6H14O3
InChI KeyXXJWXESWEXIICW-UHFFFAOYSA-N
SMILESCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Refractive Index1.4250-1.4290 @ 20?C
FormLiquid
View more
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
RUO – Research Use Only
General References:
  1. Kirk S. Schanze.; David G. Whitten. Solubilization in surfactant media: the use of an isomerizable solute-probe to determine microheterogeneity in microemulsions. J. Am. Chem. Soc. 1983, 105, (22), 6734-6735.
  2. Jacqueline T. Bork.; Jae Wook Lee.; Sonya M Khersonsky.; Ho-Sang Moon.; Young-Tae Chang. Novel Orthogonal Strategy toward Solid-Phase Synthesis of 1,3,5-Substituted Triazines . Org. Lett. 2003, 5, (2), 117-120.