Tetraethoxysilane, 98%, Thermo Scientific Chemicals
Tetraethoxysilane, 98%, Thermo Scientific Chemicals
Tetraethoxysilane, 98%, Thermo Scientific Chemicals
Tetraethoxysilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraethoxysilane, 98%, Thermo Scientific Chemicals
CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA14965.AP
Price (MYR)
436.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Specifications
Chemical Name or MaterialTetraethoxysilane
CAS78-10-4
Health Hazard 1H226-H319-H332-H335
Health Hazard 2GHS H Statement
H226-H332-H319-H335
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
View more
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
Solubility
Soluble in ethanol and 2-propanol.
Notes
Degrades in water. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Kim, S.; Ando, S.; Wang, X. Ternary composites of linear and hyperbranched polyimides with nanoscale silica for low dielectric constant, high transparency, and high thermal stability. RSC Adv. 2015, 5 (50), 40046-40054.
  2. Trogadas, P.; Parrondo, J.; Mijangos, F.; Ramani, V. Degradation mitigation in PEM fuel cells using metal nanoparticle additives. J. Mater. Chem. 2011, 21 (48), 19381-19388.